Merck
  • RNA-dependent chromatin targeting of TET2 for endogenous retrovirus control in pluripotent stem cells.
RNA-dependent chromatin targeting of TET2 for endogenous retrovirus control in pluripotent stem cells.
Nature genetics (2018-02-28)
Diana Guallar, Xianju Bi, Jose Angel Pardavila, Xin Huang, Carmen Saenz, Xianle Shi, Hongwei Zhou, Francesco Faiola, Junjun Ding, Phensinee Haruehanroengra, Fan Yang, Dan Li, Carlos Sanchez-Priego, Arven Saunders, Feng Pan, Victor Julian Valdes, Kevin Kelley, Miguel G Blanco, Lingyi Chen, Huayan Wang, Jia Sheng, Mingjiang Xu, Miguel Fidalgo, Xiaohua Shen, Jianlong Wang
ABSTRACT
Ten-eleven translocation (TET) proteins play key roles in the regulation of DNA-methylation status by oxidizing 5-methylcytosine (5mC) to generate 5-hydroxymethylcytosine (5hmC), which can both serve as a stable epigenetic mark and participate in active demethylation. Unlike the other members of the TET family, TET2 does not contain a DNA-binding domain, and it remains unclear how it is recruited to chromatin. Here we show that TET2 is recruited by the RNA-binding protein Paraspeckle component 1 (PSPC1) through transcriptionally active loci, including endogenous retroviruses (ERVs) whose long terminal repeats (LTRs) have been co-opted by mammalian genomes as stage- and tissue-specific transcriptional regulatory modules. We found that PSPC1 and TET2 contribute to ERVL and ERVL-associated gene regulation by both transcriptional repression via histone deacetylases and post-transcriptional destabilization of RNAs through 5hmC modification. Our findings provide evidence for a functional role of transcriptionally active ERVs as specific docking sites for RNA epigenetic modulation and gene regulation.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
IgG, Rabbit, Rabbit IgG Polyclonal Antibody validated for use in ELISA.