Merck
  • GABAergic signaling linked to autophagy enhances host protection against intracellular bacterial infections.
GABAergic signaling linked to autophagy enhances host protection against intracellular bacterial infections.
Nature communications (2018-10-12)
Jin Kyung Kim, Yi Sak Kim, Hye-Mi Lee, Hyo Sun Jin, Chiranjivi Neupane, Sup Kim, Sang-Hee Lee, Jung-Joon Min, Miwa Sasai, Jae-Ho Jeong, Seong-Kyu Choe, Jin-Man Kim, Masahiro Yamamoto, Hyon E Choy, Jin Bong Park, Eun-Kyeong Jo
ABSTRACT
Gamma-aminobutyric acid (GABA) is the principal inhibitory neurotransmitter in the brain; however, the roles of GABA in antimicrobial host defenses are largely unknown. Here we demonstrate that GABAergic activation enhances antimicrobial responses against intracellular bacterial infection. Intracellular bacterial infection decreases GABA levels in vitro in macrophages and in vivo in sera. Treatment of macrophages with GABA or GABAergic drugs promotes autophagy activation, enhances phagosomal maturation and antimicrobial responses against mycobacterial infection. In macrophages, the GABAergic defense is mediated via macrophage type A GABA receptor (GABAAR), intracellular calcium release, and the GABA type A receptor-associated protein-like 1 (GABARAPL1; an Atg8 homolog). Finally, GABAergic inhibition increases bacterial loads in mice and zebrafish in vivo, suggesting that the GABAergic defense plays an essential function in metazoan host defenses. Our study identified a previously unappreciated role for GABAergic signaling in linking antibacterial autophagy to enhance host innate defense against intracellular bacterial infection.
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
Muscimol, powder
Sigma-Aldrich
Kanamycin sulfate, mixture of Kanamycin A (main component) and Kanamycin B and C
Sigma-Aldrich
Isoguvacine hydrochloride, solid
Sigma-Aldrich
1(S),9(R)-(−)-Bicuculline methchloride, ≥97% (HPLC), powder
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
DAPI, for nucleic acid staining