Merck
  • Increased contractility of diabetic rabbit corpora smooth muscle in response to endothelin is mediated via Rho-kinase beta.
Increased contractility of diabetic rabbit corpora smooth muscle in response to endothelin is mediated via Rho-kinase beta.
International journal of impotence research (2003-02-28)
S Chang, J A Hypolite, A Changolkar, A J Wein, S Chacko, M E DiSanto
ABSTRACT
Corpus cavernosum smooth muscle (CCSM) from rabbits made diabetic for 6 months as a result of alloxan injection exhibited increased sensitivity (3vs 9 nM EC(50)) and generated 20-50% greater force to endothelin-1 (ET-1) compared to CCSM from normal rabbits. In contrast, the force produced by the CCSM in response to KCl and phenylephrine was not significantly altered in diabetic CCSM. The increased ET-1 sensitivity is associated with a two to three-fold upregulation of ET receptor A at both mRNA and protein levels in diabetic CCSM. ET-1-induced CCSM contraction is largely dependent upon Rho-kinase (ROK), since it is almost completely blocked by Y-27632 (a highly selective ROK inhibitor). Furthermore, expression of ROKbeta isoform is selectively upregulated in CCSM from diabetic rabbits. Thus, an increased CCSM tone, modulated by sensitization of the endothelin-mediated contractile pathway via ROK, may be a key component of the molecular mechanism of diabetes-induced erectile dysfunction.