Merck
  • DJ-1 deficiency triggers microglia sensitivity to dopamine toward a pro-inflammatory phenotype that is attenuated by rasagiline.
DJ-1 deficiency triggers microglia sensitivity to dopamine toward a pro-inflammatory phenotype that is attenuated by rasagiline.
Journal of neurochemistry (2013-12-21)
Dorit Trudler, Orly Weinreb, Silvia A Mandel, Moussa B H Youdim, Dan Frenkel
ABSTRACT
DJ-1 is an oxidative stress sensor that localizes to the mitochondria when the cell is exposed to oxidative stress. DJ-1 mutations that result in gene deficiency are linked to increased risk of Parkinson's disease (PD). Activation of microglial stress conditions that are linked to PD may result in neuronal death. We postulated that DJ-1 deficiency may increase microglial neurotoxicity. We found that down-regulation of DJ-1 in microglia using an shRNA approach increased cell sensitivity to dopamine as measured by secreted pro-inflammatory cytokines such as IL-1β and IL-6. Furthermore, we discovered that DJ-1-deficient microglia had increased monoamine oxidase activity that resulted in elevation of intracellular reactive oxygen species and nitric oxide leading to increased dopaminergic neurotoxicity. Rasagaline, a monoamine oxidase inhibitor approved for treatment of PD, reduced the microglial pro-inflammatory phenotype and significantly reduced neurotoxicity. Moreover, we discovered that DJ-1-deficient microglia have reduced expression of triggering receptor expressed on myeloid cells 2 (TREM2), previously suggested as a risk factor for pro-inflammation in neurodegenerative diseases. Further studies of DJ-1-mediated cellular pathways in microglia may contribute useful insights into the development of PD providing future avenues for therapeutic intervention
Sigma-Aldrich
MTT Formazan, powder
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture