Merck
  • Potential anti-vitiligo properties of cynarine extracted from Vernonia anthelmintica (L.) Willd.
Potential anti-vitiligo properties of cynarine extracted from Vernonia anthelmintica (L.) Willd.
International journal of molecular medicine (2018-09-19)
Nuramina Mamat, Xue Ying Lu, Maidina Kabas, Haji Akber Aisa
ABSTRACT
Vitiligo is a depigmentation disorder of the skin. It is primarily caused by the destruction of melanocytes or obstruction of the melanin synthesis pathway. Melanin is a type of skin pigment that determines skin color. The seeds of Vernonia anthelmintica (L.) Willd (Kaliziri) are used for treating skin diseases including vitiligo in traditional Uyghur medicine. 1,5‑Dicaffeoylquinic acid (1,5‑diCQA) is a natural polyphenolic compound widely distributed in plants and extracted from Kaliziri seeds. Therefore, in the present study, the effect of 1,5‑diCQA on melanin synthesis in B16 cell was evaluated, and its molecular mechanism was explored. The results indicated that 1,5‑diCQA treatment of B16 cells stimulated an increase of intracellular melanin level and tyrosinase (TYR) activity without cytotoxicity. Reverse transcription quantitative polymerase chain reaction results also indicated that 1,5‑diCQA may markedly improve the protein expression and RNA transcription of microphthalmia‑associated transcription factor (MITF), melanogenic enzyme Tyr, tyrosinase‑related protein 1 (TRP 1) and tyrosinase‑related protein 2 (TRP 2). Additional results identified that 1,5‑diCQA may promote the phosphorylation of p38 mitogen‑activated protein kinase (p38 MAPK) and extracellular signal‑regulated kinase (ERK) MAPK. Notably, the increased levels of intracellular melanin synthesis and tyrosinase expression induced by 1,5‑diCQA treatment were significantly attenuated by the protein kinase A (PKA) inhibitor H‑89. Intracellular cyclic adenosine monophosphate (cAMP) concentration and phosphorylation of cAMP‑response element binding protein was increased following 1,5‑diCQA treatment. These results indicated that 1,5‑diCQA stimulated melanogenesis via the MAPK and cAMP/PKA signaling pathways in B16 cells, which has potential therapeutic implications for vitiligo.
Product Description
Supelco
8-Methoxypsoralen, analytical standard