Merck
Targeting Fat: Mechanisms of Protein Localization to Lipid Droplets.
Trends in cell biology (2016-03-21)
Nora Kory, Robert V Farese, Tobias C Walther
ABSTRACT
How proteins specifically localize to the phospholipid monolayer surface of lipid droplets (LDs) is being unraveled. We review here the major known pathways of protein targeting to LDs and suggest a classification framework based on the localization origin for the protein. Class I proteins often have a membrane-embedded, hydrophobic 'hairpin' motif, and access LDs from the endoplasmic reticulum (ER) either during LD formation or after formation via ER-LD membrane bridges. Class II proteins access the LD surface from the cytosol and bind through amphipathic helices or other hydrophobic domains. Other proteins require lipid modifications or protein-protein interactions to bind to LDs. We summarize knowledge for targeting and removal of the different classes, and highlight areas needing investigation.
Product Description
Avanti
C4 TopFluor MG, Avanti Research - A Croda Brand 810293P, powder
Avanti
C11 TopFluor MG, Avanti Research - A Croda Brand 810296P, powder
Avanti
18:1-6:0 DNP-C11 TopFluor TG, Avanti Research - A Croda Brand 810272P, powder
Avanti
18:1-C11 TopFluor-C11 TopFluor TG, Avanti Research - A Croda Brand 810271P, powder
Avanti
18:1-18:1-C11 TopFluor TG, Avanti Research - A Croda Brand 810298C
Avanti
18:1-C11 TopFluor DG, Avanti Research - A Croda Brand 810297P, powder