Merck
  • Photodynamic Effect of Methylene Blue and Low Level Laser Radiation in Head and Neck Squamous Cell Carcinoma Cell Lines.
Photodynamic Effect of Methylene Blue and Low Level Laser Radiation in Head and Neck Squamous Cell Carcinoma Cell Lines.
International journal of molecular sciences (2018-04-13)
Barbara Kofler, Angela Romani, Christian Pritz, Teresa Bernadette Steinbichler, Volker Hans Schartinger, Herbert Riechelmann, Jozsef Dudas
ABSTRACT
Photodynamic therapy (PDT) is suggested to have an impact on the treatment of early stage head and neck cancers (HNSCC). We investigated the effect of PDT with methylene blue (MB) and a diode laser (660 nm) as the laser source on HNSCC cell lines as an in vitro model of surface oral squamous cell carcinoma. Cell-cultures were exposed to 160 µM MB for 4 min and to laser light for 8 min. Viability was proven via cell viability assay and clonogenic survival via clone counting assay. The combination of MB and diode laser evidenced high efficient loss of cell viability by 5% of the control, while treatment with the same concentration of MB for 4 min alone showed a viability of 46% of the control. In both SCC-25 and Detroit 562 HNSCC cells, MB combined with the laser allowed a significant abrogation of clonogenic growth (p < 0.01), especially in the case of Detroit 562 cells less than 1% of the suspension plated cells were able to grow tumor cell nests. Multiresistant (Detroit 562) HNSCC cells expressing cancer stem cell markers are sensitive to MB/red laser combined PDT.