Merck
  • Control of basement membrane remodeling and epithelial branching morphogenesis in embryonic lung by Rho and cytoskeletal tension.
Control of basement membrane remodeling and epithelial branching morphogenesis in embryonic lung by Rho and cytoskeletal tension.
Developmental dynamics : an official publication of the American Association of Anatomists (2004-12-23)
Kimberly A Moore, Tom Polte, Sui Huang, Bin Shi, Eben Alsberg, Mary E Sunday, Donald E Ingber
ABSTRACT
Local alterations in the mechanical compliance of the basement membrane that alter the level of isometric tension in the cell have been postulated to influence tissue morphogenesis. To explore whether cell tension contributes to tissue pattern formation in vivo, we modulated cytoskeletal force generation in embryonic mouse lung (embryonic days 12-14) rudiments using inhibitors of Rho-associated kinase (ROCK), myosin light chain kinase, myosin ATPase, and microfilament integrity, or a Rho stimulator (cytotoxic necrotizing factor-1). Tension inhibition resulted in loss of normal differentials in basement membrane thickness, inhibition of new terminal bud formation, and disorganization of epithelial growth patterns as well as disruption of capillary blood vessels. In contrast, increasing cell tension through Rho activation, as confirmed by quantitation of myosin light chain phosphorylation and immunohistocytochemical analysis of actin organization, accelerated lung branching and increase capillary elongation. These data suggest that changes in cytoskeletal tension mediated by Rho signaling through ROCK may play an important role in the establishment of the spatial differentials in cell growth and extracellular matrix remodeling that drive embryonic lung development.
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution