Merck
  • Loss of PICH Results in Chromosomal Instability, p53 Activation, and Embryonic Lethality.
Loss of PICH Results in Chromosomal Instability, p53 Activation, and Embryonic Lethality.
Cell reports (2018-09-21)
Eliene Albers, Mauro Sbroggiò, David Pladevall-Morera, Anna H Bizard, Alexandra Avram, Patricia Gonzalez, Javier Martin-Gonzalez, Ian D Hickson, Andres J Lopez-Contreras
ABSTRACT
PICH is a DNA translocase necessary for the resolution of ultrafine anaphase DNA bridges and to ensure the fidelity of chromosomal segregation. Here, we report the generation of an animal model deficient for PICH that allowed us to investigate its physiological relevance. Pich KO mice lose viability during embryonic development due to a global accumulation of DNA damage. However, despite the presence of chromosomal instability, extensive p53 activation, and increased apoptosis throughout the embryo, Pich KO embryos survive until day 12.5 of embryonic development. The absence of p53 failed to improve the viability of the Pich KO embryos, suggesting that the observed developmental defects are not solely due to p53-induced apoptosis. Moreover, Pich-deficient mouse embryonic fibroblasts exhibit chromosomal instability and are resistant to RASV12/E1A-induced transformation. Overall, our data indicate that PICH is essential to preserve chromosomal integrity in rapidly proliferating cells and is therefore critical during embryonic development and tumorigenesis.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Senescence Cells Histochemical Staining Kit, sufficient for 100 tests
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse