Merck
  • cGAS drives noncanonical-inflammasome activation in age-related macular degeneration.
cGAS drives noncanonical-inflammasome activation in age-related macular degeneration.
Nature medicine (2017-11-28)
Nagaraj Kerur, Shinichi Fukuda, Daipayan Banerjee, Younghee Kim, Dongxu Fu, Ivana Apicella, Akhil Varshney, Reo Yasuma, Benjamin J Fowler, Elmira Baghdasaryan, Kenneth M Marion, Xiwen Huang, Tetsuhiro Yasuma, Yoshio Hirano, Vlad Serbulea, Meenakshi Ambati, Vidya L Ambati, Yuji Kajiwara, Kameshwari Ambati, Shuichiro Hirahara, Ana Bastos-Carvalho, Yuichiro Ogura, Hiroko Terasaki, Tetsuro Oshika, Kyung Bo Kim, David R Hinton, Norbert Leitinger, John C Cambier, Joseph D Buxbaum, M Cristina Kenney, S Michal Jazwinski, Hiroshi Nagai, Isao Hara, A Phillip West, Katherine A Fitzgerald, SriniVas R Sadda, Bradley D Gelfand, Jayakrishna Ambati
ABSTRACT
Geographic atrophy is a blinding form of age-related macular degeneration characterized by retinal pigmented epithelium (RPE) death; the RPE also exhibits DICER1 deficiency, resultant accumulation of endogenous Alu-retroelement RNA, and NLRP3-inflammasome activation. How the inflammasome is activated in this untreatable disease is largely unknown. Here we demonstrate that RPE degeneration in human-cell-culture and mouse models is driven by a noncanonical-inflammasome pathway that activates caspase-4 (caspase-11 in mice) and caspase-1, and requires cyclic GMP-AMP synthase (cGAS)-dependent interferon-β production and gasdermin D-dependent interleukin-18 secretion. Decreased DICER1 levels or Alu-RNA accumulation triggers cytosolic escape of mitochondrial DNA, which engages cGAS. Moreover, caspase-4, gasdermin D, interferon-β, and cGAS levels were elevated in the RPE in human eyes with geographic atrophy. Collectively, these data highlight an unexpected role of cGAS in responding to mobile-element transcripts, reveal cGAS-driven interferon signaling as a conduit for mitochondrial-damage-induced inflammasome activation, expand the immune-sensing repertoire of cGAS and caspase-4 to noninfectious human disease, and identify new potential targets for treatment of a major cause of blindness.
Sigma-Aldrich
Anti-phospho-STAT2 (Tyr689) Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.