Merck
  • Analysis of ketone-based neurosteroids by reactive low-temperature plasma mass spectrometry.
Analysis of ketone-based neurosteroids by reactive low-temperature plasma mass spectrometry.
Rapid communications in mass spectrometry : RCM (2018-05-29)
Xuelu Ding, Vannuruswamy Garikapati, Bernhard Spengler, Sven Heiles
ABSTRACT
Neurosteroids are important signalling molecules that modulate neuronal activity. Their low concentrations and low volatility make neurosteroid detection and quantification by ambient mass spectrometry challenging. Here we develop a reactive low-temperature plasma mass spectrometry (LTP-MS) method and demonstrate its potential for fast screening and quantification of neurosteroids in mouse brain. Ketone-based neurosteroids were analysed with the LTP-MS method. The plasma of the LTP was heated in order to improve the desorption efficiency of low-volatility neurosteroids. Methylamine with a concentration of 500 ppbv was employed as the reactive reagent. Neurosteroids in mouse brain tissue extracts were detected in 70 s with mass errors less than ±3 ppm due to coupling of the ion source with a high-performance mass spectrometer. Reaction between neurosteroids and methylamine, seeded into the LTP gas stream, resulted in the formation of protonated methylamine-neurosteroid adducts with 5- to 100-fold abundances, compared to [M + H]+ ions detected in non-reactive LTP-MS. The lowest detectable concentrations of neurosteroid standards were in the range of ng/mL. Concentrations of neurosteroids in male and female mouse brain extracts as determined with reactive LTP-MS were on the level of ng/g, comparable to results obtained with high-performance liquid chromatography-tandem mass spectrometry. The developed reactive LTP-MS is capable of providing sensitive identification and quantification of ketone-based neurosteroids in mouse brain extracts with minimal sample treatment, and showcases the potential of reactive LTP-MS as a tool for fast screening of neurosteroid levels in brain.
Sigma-Aldrich
Pregnenolone acetate, 99%
Sigma-Aldrich
Formestane, solid