Merck
  • Dppa2/4 Facilitate Epigenetic Remodeling during Reprogramming to Pluripotency.
Dppa2/4 Facilitate Epigenetic Remodeling during Reprogramming to Pluripotency.
Cell stem cell (2018-08-28)
Charles Hernandez, Zheng Wang, Bulat Ramazanov, Yin Tang, Sameet Mehta, Cheryl Dambrot, Yu-Wei Lee, Kaleab Tessema, Ishan Kumar, Michael Astudillo, Thomas A Neubert, Shangqin Guo, Natalia B Ivanova
ABSTRACT
As somatic cells are converted into induced pluripotent stem cells (iPSCs), their chromatin is remodeled to a pluripotent configuration with unique euchromatin-to-heterochromatin ratios, DNA methylation patterns, and enhancer and promoter status. The molecular machinery underlying this process is largely unknown. Here, we show that embryonic stem cell (ESC)-specific factors Dppa2 and Dppa4 play a key role in resetting the epigenome to a pluripotent state. They are induced in reprogramming intermediates, function as a heterodimer, and are required for efficient reprogramming of mouse and human cells. When co-expressed with Oct4, Klf4, Sox2, and Myc (OKSM) factors, Dppa2/4 yield reprogramming efficiencies that exceed 80% and accelerate reprogramming kinetics, generating iPSCs in 2 to 4 days. When bound to chromatin, Dppa2/4 initiate global chromatin decompaction via the DNA damage response pathway and contribute to downregulation of somatic genes and activation of ESC enhancers, all of which enables an efficient transition to pluripotency. Our work provides critical insights into how the epigenome is remodeled during acquisition of pluripotency.
Sigma-Aldrich
MEK1/2 Inhibitor III, PD0325901, InSolution, ≥95%
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Roche
KAPA Express Extract, with PCR ReadyMix
Sigma-Aldrich
Anti-DPPA-2 Antibody, clone 6C1.2, clone 6C1.2, from mouse