Merck
  • Cholera toxin inhibits SNX27-retromer-mediated delivery of cargo proteins to the plasma membrane.
Cholera toxin inhibits SNX27-retromer-mediated delivery of cargo proteins to the plasma membrane.
Journal of cell science (2018-07-22)
Varsha Singh, Jianbo Yang, Jianyi Yin, Robert Cole, Ming Tse, Diego E Berman, Scott A Small, Gregory Petsko, Mark Donowitz
ABSTRACT
Cholera toxin (CT) causes severe diarrhea by increasing intracellular cAMP leading to a PKA-dependent increase in Cl- secretion through CFTR and decreased Na+ absorption through inhibition of Na+/H+ exchanger 3 (NHE3; also known as SLC9A3). The mechanism(s) by which CT inhibits NHE3 is partially understood, although no drug therapy has been successful at reversing this inhibition. We now describe that CT phosphorylates an amino acid in the PDZ domain of SNX27, which inhibits SNX27-mediated trafficking of NHE3 from the early endosomes to the plasma membrane (PM), and contributes to reduced basal NHE3 activity through a mechanism that involves reduced PM expression and reduced endocytic recycling. Importantly, mutagenesis studies (Ser to Asp) showed that the effect of this phosphorylation of SNX27 phenocopies the effects seen upon loss of SNX27 function, affecting PM trafficking of cargo proteins that bind SNX27-retromer. Additionally, CT destabilizes retromer function by decreasing the amount of core retromer proteins. These effects of CT can be partially rescued by enhancing retromer stability by using 'pharmacological chaperones'. Moreover, pharmacological chaperones can be used to increase basal and cholera toxin-inhibited NHE3 activity and fluid absorption by intestinal epithelial cells.This article has an associated First Person interview with the first author of the paper.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.