Merck
  • The ATPase VCP/p97 functions as a disaggregase against toxic Huntingtin-exon1 aggregates.
The ATPase VCP/p97 functions as a disaggregase against toxic Huntingtin-exon1 aggregates.
FEBS letters (2018-08-03)
Debasish Kumar Ghosh, Ajit Roy, Akash Ranjan
ABSTRACT
Intracellular protein aggregation is characterized by accumulation of misfolded proteins. Chaperones, degradation machineries, and quality-control mechanisms counteract protein aggregation. In this study, we report that the ATPase valosin-containing protein (VCP/p97) acts as a functional disaggregase that disassembles Huntingtin-exon1 aggregates in vitro and in HeLa cells. The N-terminal part of VCP (Cdc48_N domain) interacts with the N-terminal 17-amino acid region of Huntingtin-exon1. We show that VCP has properties of a disaggregase, since it is capable of reducing preformed protein aggregates and displays increased ATPase activity in the presence of protein aggregates. However, VCP shows high divergence/disparity from other disaggregases. Taken together, our studies show the novel function of VCP/p97 as a disaggregase which detangles protein aggregates to probably channelize their degradation.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Supelco
Malachite Green chloride, analytical standard
Sigma-Aldrich
Malachite Green Phosphate Assay Kit, sufficient for 2500 colorimetric tests