Merck
  • Defects in B lymphocyte maturation and T lymphocyte activation in mice lacking Jak3.
Defects in B lymphocyte maturation and T lymphocyte activation in mice lacking Jak3.
Science (New York, N.Y.) (1995-11-03)
D C Thomis, C B Gurniak, E Tivol, A H Sharpe, L J Berg
ABSTRACT
Biochemical studies of signaling mediated by many cytokine and growth factor receptors have implicated members of the Jak family of tyrosine kinases in these pathways. Specifically, Jak3 has been shown to be associated with the interleukin-2 (IL-2) receptor gamma chain, a component of the receptors for IL-2, IL-4, IL-7, IL-9, and IL-15. Mice lacking Jak3 showed a severe block in B cell development at the pre-B stage in the bone marrow. In contrast, although the thymuses of these mice were small, T cell maturation progressed relatively normally. In response to mitogenic signals, peripheral T cells in Jak3-deficient mice did not proliferate and secreted small amounts of IL-2. These data demonstrate that Jak3 is critical for the progression of B cell development in the bone marrow and for the functional competence of mature T cells.