Merck
Protein-tyrosine phosphatase expression in pre-B cell NALM-6.
Cancer research (1992-02-01)
M Adachi, M Sekiya, Y Arimura, M Takekawa, F Itoh, Y Hinoda, K Imai, A Yachi
ABSTRACT
Protein-tyrosine phosphatase (PTP)-related complementary DNAs from NALM-6 (pre-B cell line) were amplified by reverse transcriptase polymerase chain reaction using primers corresponding to the conserved catalytic domains of PTPs. Thirty-three polymerase chain reaction products, identified as PTP related complementary DNAs, were classified to RPTP-alpha, PTP1B, and 4 novel PTPs, which were designated as BPTP-1-4. Their expressions in NALM-6 and other cell lines were confirmed by Northern blot analysis. BPTP-1 and -2 exhibited extensive homology with the first and the second catalytic domains, respectively, of leukocyte common antigen related molecule (LAR) and human PTP delta. The transcriptional sizes of BPTP-1 and BPTP-2 are the same (7.2 kilobases) as that of LAR. The expression of BPTP-1 was abundant in lymphoid cell lines TALL-1 and NALM-6 but small in colon cell line BM314, which is in sharp contrast to the expression of LAR. These data suggest that the expression levels of BPTP-1 and LAR are altered in a cell specific manner, probably making them cell type associated PTPs.