Merck
  • SH2B1 modulates chromatin state and MyoD occupancy to enhance expressions of myogenic genes.
SH2B1 modulates chromatin state and MyoD occupancy to enhance expressions of myogenic genes.
Biochimica et biophysica acta. Gene regulatory mechanisms (2017-01-01)
Kuan-Wei Chen, Yu-Jung Chang, Chia-Ming Yeh, Yen-Ling Lian, Michael W Y Chan, Cheng-Fu Kao, Linyi Chen
ABSTRACT
As mesoderm-derived cell lineage commits to myogenesis, a spectrum of signaling molecules, including insulin growth factor (IGF), activate signaling pathways and ultimately instruct chromatin remodeling and the transcription of myogenic genes. MyoD is a key transcription factor during myogenesis. In this study, we have identified and characterized a novel myogenic regulator, SH2B1. Knocking down SH2B1 delays global chromatin condensation and decreases the formation of myotubes. SH2B1 interacts with histone H1 and is required for the removal of histone H1 from active transcription sites, allowing for the expressions of myogenic genes, IGF2 and MYOG. Chromatin immunoprecipitation assays suggest the requirement of SH2B1 for the induction of histone H3 lysine 4 trimethylation as well as the reduction of histone H3 lysine 9 trimethylation at the promoters and/or enhancers of IGF2 and MYOG genes during myogenesis. Furthermore, SH2B1 is required for the transcriptional activity of MyoD and MyoD occupancy at the enhancer/promoter regions of IGF2 and MYOG during myogenesis. Together, this study demonstrates that SH2B1 fine-tunes global-local chromatin states, expressions of myogenic genes and ultimately promotes myogenesis.
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI