Merck
  • Activated EphA2 Processing by MT1-MMP Is Involved in Malignant Transformation of Ovarian Tumours In Vivo.
Activated EphA2 Processing by MT1-MMP Is Involved in Malignant Transformation of Ovarian Tumours In Vivo.
Anticancer research (2018-07-05)
Yoko Takahashi, Makoto Hamasaki, Mikiko Aoki, Kaori Koga, Naohiko Koshikawa, Shingo Miyamoto, Kazuki Nabeshima
ABSTRACT
Erythropoietin-producing hepatocellular receptor-2 (EphA2) is overexpressed in ovarian cancer. The N-terminals of EphA2 are processed by membrane-type 1 matrix metalloproteinase (MT1-MMP) and can subsequently induce ligand-independent signal activation to promote motility, invasion, and metastasis. The aim of this study was to investigate whether EphA2 processing occurs in benign, borderline, and malignant ovarian tumours. Overall 107 ovarian epithelial carcinomas (OECs; 47 serous, 24 endometrioid, 16 mucinous, and 20 clear cell), 54 ovarian borderline tumours (OBTs; 12 serous, 42 mucinous), and 45 adenomas (15 serous, 17 mucinous, and 13 endometriotic cysts) were evaluated. Expression and processing of EphA2 were semi-quantitatively analyzed. EphA2 processing was also investigated by immunoblotting. EphA2 and MT1-MMP co-expression were detected. N-terminal EphA2 levels were significantly lower than those of C-terminal EphA2 in OECs and OBTs, but not in adenomas. Immunoblotting revealed processed fragments in OEC and OBTs. EphA2 processing by MT1-MMP is associated with malignant transformation in ovarian tumours.
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block