Merck
  • Metabolic enzyme PFKFB4 activates transcriptional coactivator SRC-3 to drive breast cancer.
Metabolic enzyme PFKFB4 activates transcriptional coactivator SRC-3 to drive breast cancer.
Nature (2018-04-05)
Subhamoy Dasgupta, Kimal Rajapakshe, Bokai Zhu, Bryan C Nikolai, Ping Yi, Nagireddy Putluri, Jong Min Choi, Sung Y Jung, Cristian Coarfa, Thomas F Westbrook, Xiang H-F Zhang, Charles E Foulds, Sophia Y Tsai, Ming-Jer Tsai, Bert W O'Malley
ABSTRACT
Alterations in both cell metabolism and transcriptional programs are hallmarks of cancer that sustain rapid proliferation and metastasis 1 . However, the mechanisms that control the interaction between metabolic reprogramming and transcriptional regulation remain unclear. Here we show that the metabolic enzyme 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 4 (PFKFB4) regulates transcriptional reprogramming by activating the oncogenic steroid receptor coactivator-3 (SRC-3). We used a kinome-wide RNA interference-based screening method to identify potential kinases that modulate the intrinsic SRC-3 transcriptional response. PFKFB4, a regulatory enzyme that synthesizes a potent stimulator of glycolysis 2 , is found to be a robust stimulator of SRC-3 that coregulates oestrogen receptor. PFKFB4 phosphorylates SRC-3 at serine 857 and enhances its transcriptional activity, whereas either suppression of PFKFB4 or ectopic expression of a phosphorylation-deficient Ser857Ala mutant SRC-3 abolishes the SRC-3-mediated transcriptional output. Functionally, PFKFB4-driven SRC-3 activation drives glucose flux towards the pentose phosphate pathway and enables purine synthesis by transcriptionally upregulating the expression of the enzyme transketolase. In addition, the two enzymes adenosine monophosphate deaminase-1 (AMPD1) and xanthine dehydrogenase (XDH), which are involved in purine metabolism, were identified as SRC-3 targets that may or may not be directly involved in purine synthesis. Mechanistically, phosphorylation of SRC-3 at Ser857 increases its interaction with the transcription factor ATF4 by stabilizing the recruitment of SRC-3 and ATF4 to target gene promoters. Ablation of SRC-3 or PFKFB4 suppresses breast tumour growth in mice and prevents metastasis to the lung from an orthotopic setting, as does Ser857Ala-mutant SRC-3. PFKFB4 and phosphorylated SRC-3 levels are increased and correlate in oestrogen receptor-positive tumours, whereas, in patients with the basal subtype, PFKFB4 and SRC-3 drive a common protein signature that correlates with the poor survival of patients with breast cancer. These findings suggest that the Warburg pathway enzyme PFKFB4 acts as a molecular fulcrum that couples sugar metabolism to transcriptional activation by stimulating SRC-3 to promote aggressive metastatic tumours.
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Green, Duolink® PLA kit for Flow Cytometry with Green Detection
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
HUF 75,800.00
50 μg
Please contact Customer Service for Availability
HUF 135,000.00