Merck
  • 3-substitued indoles: one-pot synthesis and evaluation of anticancer and Src kinase inhibitory activities.
3-substitued indoles: one-pot synthesis and evaluation of anticancer and Src kinase inhibitory activities.
Bioorganic & medicinal chemistry letters (2011-05-27)
V Kameshwara Rao, Bhupender S Chhikara, Amir Nasrolahi Shirazi, Rakesh Tiwari, Keykavous Parang, Anil Kumar
ABSTRACT
An efficient and economical method was developed for the synthesis of 3-substituted indoles by one-pot three-component coupling reaction of a substituted or unsubstituted benzaldehyde, N-methylaniline, and indole or N-methylindole using Yb(OTf)(3)-SiO(2) as a catalyst. All the synthesized compounds were evaluated for inhibition of cell proliferation of human colon carcinoma (HT-29), human ovarian adenocarcinoma (SK-OV-3), and c-Src kinase activity. The 4-methylphenyl (4o and 4p) and 4-methoxyphenyl (4q) indole derivatives inhibited the cell proliferation of SK-OV-3 and HT-29 cells by 70-77% at a concentration of 50 μM. The unsubstituted phenyl (4d) and 3-nitrophenyl (4l) derivatives showed the inhibition of c-Src kinase with IC(50) values of 50.6 and 58.3 μM, respectively.
Sigma-Aldrich
N-Methylaniline, purum, ≥98.0% (GC)
Supelco
N-Methylaniline, analytical standard
Sigma-Aldrich
N-Methylaniline, ≥99%
Sigma-Aldrich
N-Methylaniline, 98%