Merck
Styrene in foods and dietary exposure estimates.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2018-08-28)
Xu-Liang Cao, Melissa Sparling, Luc Pelletier, Robert Dabeka
ABSTRACT
Low levels of styrene may be found in foods as a result of possible migration from polystyrene-based food packaging and as a result of its formation during the biodegradation of a wide variety of naturally occurring compounds with structures similar to styrene. In this study, composite food samples from a recent (2014) Canadian Total Diet Study were analysed for styrene, and levels of styrene in samples of most food types were low in general with a few exceptions (e.g. 4934 ng/g in herbs and spices). Dietary exposures to styrene were estimated for different age-groups based on the occurrence data and the food consumption data for all persons, and they are 0.17-0.38 µg/kg body weight/day for children and 0.12-0.16 µg/kg body weight/day for adults, similar to air intakes (0.085-0.27 µg/kg body weight/day). Thus, for the general population, both food and air contribute similar portions of the total daily intake of styrene for all age groups. However, for the smoking population, intakes from cigarettes are still the major route of exposure to styrene.
Product Description
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, metal alloy fiber, for use with manual holder
Pricing and availability is not currently available.
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME Portable Field Sampler, coating PDMS
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Pricing and availability is not currently available.
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Pricing and availability is not currently available.
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub
Pricing and availability is not currently available.
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Pricing and availability is not currently available.
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Pricing and availability is not currently available.
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Pricing and availability is not currently available.
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder
Pricing and availability is not currently available.
Supelco
SPME Sampling Stand, for use with 4 mL vials
Pricing and availability is not currently available.
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with manual holder
Pricing and availability is not currently available.
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
Pricing and availability is not currently available.
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Pricing and availability is not currently available.
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Pricing and availability is not currently available.
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga
Pricing and availability is not currently available.
Supelco
SPME Sampling Stand, for use with 15 mL vials
Pricing and availability is not currently available.