Merck
  • Inhibition of ovarian tumor cell invasiveness by targeting SYK in the tyrosine kinase signaling pathway.
Inhibition of ovarian tumor cell invasiveness by targeting SYK in the tyrosine kinase signaling pathway.
Oncogene (2018-04-13)
Yu Yu, Yohan Suryo Rahmanto, Meng-Horng Lee, Pei-Hsun Wu, Jude M Phillip, Chuan-Hsiang Huang, Michele I Vitolo, Stephanie Gaillard, Stuart S Martin, Denis Wirtz, Ie-Ming Shih, Tian-Li Wang
ABSTRACT
Cell motility and invasiveness are prerequisites for dissemination, and largely account for cancer mortality. We have identified an actionable kinase, spleen tyrosine kinase (SYK), which is keenly tightly associated with tumor progression in ovarian cancer. Here, we report that active recombinant SYK directly phosphorylates cortactin and cofilin, which are critically involved in assembly and dynamics of actin filament through phosphorylation signaling. Enhancing SYK activity by inducing expression of a constitutively active SYK mutant, SYK130E, increased growth factor-stimulated migration and invasion of ovarian cancer cells, which was abrogated by cortactin knockdown. Similarly, SYK inhibitors significantly decreased invasion of ovarian cancer cells across basement membrane in real-time transwell assays and in 3D tumor spheroid models. SYK inactivation by targeted gene knockout or by small molecule inhibition reduced actin polymerization. Collectively, this study reported a new mechanism by which SYK signaling regulates ovarian cancer cell motility and invasiveness, and suggest a target-based strategy to prevent or suppress the advancement of ovarian malignancies.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-Cortactin Antibody, phospho-specific (Tyr421), Chemicon®, from rabbit