Merck
  • Pannexin-1 in Human Lymphatic Endothelial Cells Regulates Lymphangiogenesis.
Pannexin-1 in Human Lymphatic Endothelial Cells Regulates Lymphangiogenesis.
International journal of molecular sciences (2018-06-09)
Jonathan Boucher, Claire Simonneau, Golthlay Denet, Jonathan Clarhaut, Annie-Claire Balandre, Marc Mesnil, Laurent Cronier, Arnaud Monvoisin
ABSTRACT
The molecular mechanisms governing the formation of lymphatic vasculature are not yet well understood. Pannexins are transmembrane proteins that form channels which allow for diffusion of ions and small molecules (<1 kDa) between the extracellular space and the cytosol. The expression and function of pannexins in blood vessels have been studied in the last few decades. Meanwhile, no studies have been conducted to evaluate the role of pannexins during human lymphatic vessel formation. Here we show, using primary human dermal lymphatic endothelial cells (HDLECs), pharmacological tools (probenecid, Brilliant Blue FCF, mimetic peptides [10Panx]) and siRNA-mediated knockdown that Pannexin-1 is necessary for capillary tube formation on Matrigel and for VEGF-C-induced invasion. These results newly identify Pannexin-1 as a protein highly expressed in HDLECs and its requirement during in vitro lymphangiogenesis.
Sigma-Aldrich
MISSION® esiRNA, targeting human PANX1
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody