Merck
  • Chemogenetic control of gene expression and cell signaling with antiviral drugs.
Chemogenetic control of gene expression and cell signaling with antiviral drugs.
Nature methods (2018-07-04)
Elliot P Tague, Hannah L Dotson, Shannon N Tunney, D Christopher Sloas, John T Ngo
ABSTRACT
We developed a method in which the NS3 cis-protease from hepatitis C virus can be used as a ligand-inducible connection to control the function and localization of engineered proteins in mammalian cells. To demonstrate the versatility of this approach, we designed drug-sensitive transcription factors and transmembrane signaling proteins, the activities of which can be tightly and reversibly controlled through the use of clinically tested antiviral protease inhibitors.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture