Merck
  • Herpes simplex virus-1 entry into cells mediated by a novel member of the TNF/NGF receptor family.
Herpes simplex virus-1 entry into cells mediated by a novel member of the TNF/NGF receptor family.
Cell (1996-11-01)
R I Montgomery, M S Warner, B J Lum, P G Spear
ABSTRACT
We identified and cloned a cellular mediator of herpes simplex virus (HSV) entry. Hamster and swine cells resistant to viral entry became susceptible upon expression of a human cDNA encoding this protein, designated HVEM (for herpesvirus entry mediator). HVEM was shown to mediate the entry of several wild-type HSV strains of both serotypes. Anti-HVEM antibodies and a soluble hybrid protein containing the HVEM ectodomain inhibited HVEM-dependent infection but not virus binding to cells. Mutations in the HSV envelope glycoprotein gD significantly reduced HVEM-mediated entry. The contribution of HVEM to HSV entry into human cells was demonstrable in activated T cells. HVEM, the first identified mediator of HSV entry, is a new member of the TNF/NGF receptor family.
Sigma-Aldrich
CD270 human, recombinant, expressed in E. coli, 0.5 mg protein/mL