Merck
  • IRE1α RNase-dependent lipid homeostasis promotes survival in Myc-transformed cancers.
IRE1α RNase-dependent lipid homeostasis promotes survival in Myc-transformed cancers.
The Journal of clinical investigation (2018-01-31)
Hong Xie, Chih-Hang Anthony Tang, Jun H Song, Anthony Mancuso, Juan R Del Valle, Jin Cao, Yan Xiang, Chi V Dang, Roy Lan, Danielle J Sanchez, Brian Keith, Chih-Chi Andrew Hu, M Celeste Simon
ABSTRACT
Myc activation is a primary oncogenic event in many human cancers; however, these transcription factors are difficult to inhibit pharmacologically, suggesting that Myc-dependent downstream effectors may be more tractable therapeutic targets. Here, we show that Myc overexpression induces endoplasmic reticulum (ER) stress and engages the inositol-requiring enzyme 1α (IRE1α)/X-box binding protein 1 (XBP1) pathway through multiple molecular mechanisms in a variety of c-Myc- and N-Myc-dependent cancers. In particular, Myc-overexpressing cells require IRE1α/XBP1 signaling for sustained growth and survival in vitro and in vivo, dependent on elevated stearoyl-CoA-desaturase 1 (SCD1) activity. Pharmacological and genetic XBP1 inhibition induces Myc-dependent apoptosis, which is alleviated by exogenous unsaturated fatty acids. Of note, SCD1 inhibition phenocopies IRE1α RNase activity suppression in vivo. Furthermore, IRE1α inhibition enhances the cytotoxic effects of standard chemotherapy drugs used to treat c-Myc-overexpressing Burkitt's lymphoma, suggesting that inhibiting the IRE1α/XBP1 pathway is a useful general strategy for treatment of Myc-driven cancers.
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
4-Ethylmorpholine, BioXtra, suitable for protein sequencing, ≥99.5% (GC)
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
ApopTag Plus In Situ Apoptosis Fluorescein Detection Kit, The ApopTag Plus Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a fluorescein reporter molecule.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Roche
cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.