Merck
Potential treatment for vocal fold scar with pirfenidone.
The Laryngoscope (2017-11-25)
Haruka Kodama, Yoshihiko Kumai, Kohei Nishimoto, Yutaka Toya, Satoru Miyamaru, Shinobu Furushima, Eiji Yumoto
ABSTRACT
Pirfenidone (PFD) is a strong antifibrotic agent that has been clinically approved in Japan for idiopathic pulmonary fibrosis. We examined the antifibrotic effects of PFD on fibroblasts isolated from scarred vocal folds (VFs) of ferrets in vitro. Prospective animal experiments with controls. Scar fibroblasts (SFs) were isolated from scarred VFs that had been electrocauterized 2 weeks before harvesting (N = 4). Normal fibroblasts (NFs) were isolated from intact VFs (N = 4). SFs and NFs were incubated in the presence of 10 ng/mL transforming growth factor β1 (TGF-β1), with or without PFD. After the 48-hour incubation, mRNA expression levels of α smooth muscle actin (αSMA), TGF-β1, collagen type I, and hyaluronan synthase 2 (HAS2) were examined by real-time polymerase chain reaction. Immunohistochemistry with anti-αSMA anti-collagen type I and phosphorylated Smad (p-Smad)2/3 antibodies in SFs with or without PFD was performed. SFs and NFs were cultured in collagen gel with or without PFD for 48 hours, and the extent of gel contraction was examined quantitatively. PFD treatment significantly (P < .05) decreased mRNA expression of collagen type I, significantly increased mRNA expression of TGF-β1 and HAS2, and significantly suppressed collagen gel contraction. However, it did not have a significant effect on the expression of αSMA. The expression of p-Smad2/3 in the nucleus was faded with PFD, possibly demonstrating the suppression of translocation of p-Smad2/3 from cytoplasm to nucleus with PFD. This is the first report to demonstrate the in vitro antifibrotic effects of PFD on fibroblasts isolated from scarred VFs of ferrets. NA. Laryngoscope, 128:E171-E177, 2018.
Sigma-Aldrich
Transforming Growth Factor-β1 human, TGF-β1, recombinant, expressed in CHO cells, powder, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mL
Please contact Customer Service for Availability
NOK 950.00
500 mL
Please contact Customer Service for Availability
NOK 2,030.00
1 L
Please contact Customer Service for Availability
NOK 2,550.00