Merck
  • ELISA Development for Serum Hemeoxygenase-1 and Its Application to Patients with Acute Respiratory Distress Syndrome.
ELISA Development for Serum Hemeoxygenase-1 and Its Application to Patients with Acute Respiratory Distress Syndrome.
Canadian respiratory journal (2018-06-01)
Yu Hara, Masaharu Shinkai, Masataka Taguri, Kenjiro Nagai, Satoru Hashimoto, Takeshi Kaneko
ABSTRACT
Hemeoxygenase-1 (HO-1) is an essential enzyme in heme catabolism and has been proposed as a biomarker of lung disease prognosis. We modified a commercial HO-1 enzyme-linked immunosorbent assay (ELISA) kit to achieve higher sensitivity and evaluated if serum HO-1 could be a biomarker to predict the prognosis of acute respiratory distress syndrome (ARDS) patients. Serum samples were collected from 15 healthy volunteers to validate the modified ELISA. In the 22 patients with ARDS who were enrolled, serum HO-1 was measured upon diagnosis (D0) and at 7 days after diagnosis (D7). The serum HO-1 concentration could be measured in all healthy volunteers. The intra- and interassay tests and the percentage recovery test were acceptable. Compared with normal control subjects, patients with ARDS had significantly higher D0 HO-1 concentrations (75.4 ng/mL versus 31.7 ng/mL, P < 0.001). The 28-day survival was significantly better in patients with low D0 HO-1 (<75.8 ng/mL) than in those with high D0 HO-1 (≥75.8 ng/mL) (mortality rate: 18% versus 73%, P=0.016). Nonsurvivors had significantly higher D0 and D7 HO-1 concentrations than survivors (P < 0.05). Serum HO-1 may be a useful biomarker to predict the prognosis of patients with ARDS.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%