Merck
  • Cold-inducible RNA-binding protein (CIRP) induces translation of the cell-cycle inhibitor p27Kip1.
Cold-inducible RNA-binding protein (CIRP) induces translation of the cell-cycle inhibitor p27Kip1.
Nucleic acids research (2018-01-24)
Martina Roilo, Michael K Kullmann, Ludger Hengst
ABSTRACT
The CDK inhibitor p27Kip1 plays a central role in controlling cell proliferation and cell-cycle exit. p27Kip1 protein levels oscillate during cell-cycle progression and are regulated by mitogen or anti-proliferative signaling. The abundance of the protein is frequently determined by post-transcriptional mechanisms including ubiquitin-mediated proteolysis and translational control. Here, we report that the cold-inducible RNA-binding protein (CIRP) selectively binds to the 5' untranslated region of the p27Kip1 mRNA. CIRP is induced, modified and relocalized in response to various stress stimuli and can regulate cell survival and cell proliferation particularly during stress. Binding of CIRP to the 5'UTR of the p27Kip1 mRNA significantly enhanced reporter translation. In cells exposed to mild hypothermia, the induction of CIRP correlated with increased translation of a p27Kip1 5'UTR reporter and with the accumulation of p27Kip1 protein. shRNA-mediated CIRP knockdown could prevent the induction of translation. We found that p27Kip1 is central for the decreased proliferation at lower temperature, since p27Kip1 KO mouse embryonic fibroblasts (MEFs) hardly increased their doubling time in hypothermic conditions, whereas wild-type MEFs significantly delayed proliferation in response to cold stress. This suggests that the CIRP-dependent p27Kip1 upregulation during mild hypothermia contributes to the cold shock-induced inhibition of cell proliferation.
Sigma-Aldrich
MISSION® esiRNA, targeting human CIRBP
Sigma-Aldrich
E-64d, protease inhibitor
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)