Merck
  • Top-down LC-MS quantitation of intact denatured and native monoclonal antibodies in biological samples.
Top-down LC-MS quantitation of intact denatured and native monoclonal antibodies in biological samples.
Bioanalysis (2018-06-29)
Liyun Zhang, Lisa A Vasicek, SuChun Hsieh, Shuli Zhang, Kevin P Bateman, Jack Henion
ABSTRACT
The requirements for developing antibody biotherapeutics benefit from understanding the nature and relevant aspects of the entire molecule. The method presented herein employs on-line multidimensional LC-quadrupole time-of-flight (QTOF)-MS for the quantitative determination of an antibody isolated from biological samples while maintaining the intact native biologically active conformation of the antibody. Following method optimization for a model antibody, an incurred biotherapeutic in cynomologus monkey was quantified in its intact top-down native conformation. A partial method validation demonstrated acceptable precision and accuracy although improved sensitivity requires further studies. An on-line multidimensional LC-MS approach presents a proof-of-principle example for quantifying an intact, native antibody isolated from an incurred biological sample via immunoaffinity techniques coupled with top-down QTOF LC-MS bioanalysis.
Sigma-Aldrich
SILuMAB Stable-Isotope Labeled Universal Monoclonal Antibody Standard human, recombinant, expressed in CHO cells
Sigma-Aldrich
SILuMAB K1 Stable-Isotope Labeled Universal Monoclonal Antibody, recombinant, expressed in CHO cells