Merck
  • Overexpression of suppressor of IKBKE 1 is associated with vincristine resistance in colon cancer cells.
Overexpression of suppressor of IKBKE 1 is associated with vincristine resistance in colon cancer cells.
Biomedical reports (2016-11-25)
Chun-Peng Zhao, Zhong-Jie Xu, Qing Guo, Yun-Xiao Li, Xiang-Zheng Gao, Yi-You Peng
ABSTRACT
In a previous study, the suppressor of IKBKE 1 expression level was confirmed to be higher in vincristine (VCR)-resistant HCT-8 (HCT-8/V) colon cancer cells than in non-VCR-resistant HCT-8 cells. In the current study, IKBKE 1 expression in VCR-resistant colon cancer cells was investigated further. HCT-8 and HCT-8/V human colon cancer cells were used, and polymerase chain reaction (PCR) primers were designed to amplify the IKBKE 1 gene. Fluorescence reverse transcription-quantitative PCR (RT-qPCR) was performed to detect differences in IKBKE 1 expression between sensitive and drug-resistant colon cancer cell lines. Western blotting was performed to further observe IKBKE 1 expression. Based on the RT-qPCR and western blot results, IKBKE 1 expression was observed to be markedly higher in the HCT-8/V cells, and this difference was significant (P<0.05). Thus, IKBKE 1 expression was identified to be associated with the resistance of colon cancer cells to VCR.