Merck
  • Combination therapy with an anti-IL-1β antibody and GAD65 DNA vaccine can reverse recent-onset diabetes in the RIP-GP mouse model.
Combination therapy with an anti-IL-1β antibody and GAD65 DNA vaccine can reverse recent-onset diabetes in the RIP-GP mouse model.
Diabetes (2014-02-13)
Philippe P Pagni, Damien Bresson, Teresa Rodriguez-Calvo, Amira Bel Hani, Yulia Manenkova, Natalie Amirian, Alecia Blaszczak, Sina Faton, Sowbarnika Sachithanantham, Matthias G von Herrath
ABSTRACT
Type 1 diabetes is thought to be an autoimmune condition in which self-reactive T cells attack insulin-secreting pancreatic β-cells. As a proinflammatory cytokine produced by β-cells or macrophages, interleukin-1β (IL-1β) represents a potential therapeutic target in diabetes. We reasoned IL-1β blockade could be combined with islet antigen-specific approaches involving GAD of 65 kDa (GAD65)-expressing plasmids, as previously shown in combination therapies (CTs) with anti-CD3. Thus, we investigated whether anti-IL-1β antibody alone or combined with GAD65 vaccine could reverse diabetes development in a virus-induced mouse model. Given alone, anti-IL-1β had no effect on diabetes, while GAD65 plasmid resulted in 33% disease reversal after a 5-week observation. However, CTs cured 53% of animals and prevented worsening of glycemic control in nonprotected individuals for up to 12 weeks. While the GAD65 vaccine arm of the CT was associated with increased forkhead box p3(+) regulatory T-cell frequency in pancreatic lymph nodes, islet infiltration by CD11b(+/high) cells was less frequent upon CT, and its extent correlated with treatment success or failure. Altogether, our CTs provided prolonged improvement of clinical and immunological features. Despite unsuccessful clinical trials using anti-IL-1β monotherapy, these data hold promise for treatment of type 1 diabetic patients with IL-1β blockade combined with antigen-specific vaccines.