Merck
  • Fibronectin Overexpression Modulates Formation of Macrophage Foam Cells by Activating SREBP2 Involved in Endoplasmic Reticulum Stress.
Fibronectin Overexpression Modulates Formation of Macrophage Foam Cells by Activating SREBP2 Involved in Endoplasmic Reticulum Stress.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-07-18)
Hansong Du, Yu Wang, Zhengfeng Zhang, Jing Yang, Jie Zhang, Ying Zhang
ABSTRACT
To explore the explicit role of fibronectin (FN) isforms in atherosclerotic lesions and the underlying mechanisms. Inducible stable expression was performed, and similar results were observed between EDA+FN (FN containing EDA domain) and EDA-FN (FN devoid of EDA domain). FN isforms could trigger endoplasmic reticulum (ER) stress, thereby leading to lipid accumulation in cultured Raw264.7 cells. FN isforms-induced gene expression and lipid accumulation were inhibited by a chemical chaperone 4-phenyl butyric acid (PBA) or by overexpression of the ER chaperone, GRP78/BiP, demonstrating a direct role of ER stress in activation of cholesterol/triglyceride biosynthesis. Moreover, activation of the sterol regulatory element binding protein-2 (SREBP2) was found to be downstream of ER stress, and this activation was affirmed to account for the intracellular accumulation of cholesterol using RNAi technique. our study suggests that enhanced FN in lesions facilitates foam cell formation due to dysregulation of the endogenous sterol response pathway by activation of ER stress, and confirms that EDA+FN has no more pro-atherogenic role than EDA-FN in triggering ER stress.