Merck
  • Targeting of NAT10 enhances healthspan in a mouse model of human accelerated aging syndrome.
Targeting of NAT10 enhances healthspan in a mouse model of human accelerated aging syndrome.
Nature communications (2018-04-29)
Gabriel Balmus, Delphine Larrieu, Ana C Barros, Casey Collins, Monica Abrudan, Mukerrem Demir, Nicola J Geisler, Christopher J Lelliott, Jacqueline K White, Natasha A Karp, James Atkinson, Andrea Kirton, Matt Jacobsen, Dean Clift, Raphael Rodriguez, David J Adams, Stephen P Jackson
ABSTRACT
Hutchinson-Gilford Progeria Syndrome (HGPS) is a rare, but devastating genetic disease characterized by segmental premature aging, with cardiovascular disease being the main cause of death. Cells from HGPS patients accumulate progerin, a permanently farnesylated, toxic form of Lamin A, disrupting the nuclear shape and chromatin organization, leading to DNA-damage accumulation and senescence. Therapeutic approaches targeting farnesylation or aiming to reduce progerin levels have provided only partial health improvements. Recently, we identified Remodelin, a small-molecule agent that leads to amelioration of HGPS cellular defects through inhibition of the enzyme N-acetyltransferase 10 (NAT10). Here, we show the preclinical data demonstrating that targeting NAT10 in vivo, either via chemical inhibition or genetic depletion, significantly enhances the healthspan in a Lmna G609G HGPS mouse model. Collectively, the data provided here highlights NAT10 as a potential therapeutic target for HGPS.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Sample Buffer, Laemmli 2× Concentrate
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin solution
Sigma-Aldrich
TWEEN® 80, BioXtra, viscous liquid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid