Merck
  • YB-1 regulates tumor growth by promoting MACC1/c-Met pathway in human lung adenocarcinoma.
YB-1 regulates tumor growth by promoting MACC1/c-Met pathway in human lung adenocarcinoma.
Oncotarget (2017-06-19)
Tao Guo, Shilei Zhao, Peng Wang, Xiaoyuan Xue, Yan Zhang, Mengying Yang, Nan Li, Zhuoshi Li, Lingzhi Xu, Lei Jiang, Lei Zhao, Patrick C Ma, Rafael Rosell, Jinxiu Li, Chundong Gu
ABSTRACT
Aberrant overexpression of the transcription/translation factor Y-box-binding protein (YB-1) is associated with poor prognosis of lung adenocarcinoma, however the underlying mechanism by which YB-1 acts has not been fully elucidated. Here, we reported that inhibition of YB-1 diminished proliferation, migration and invasion of lung adenocarcinoma cells. Interestingly, we identified metastasis associated in colon cancer-1 (MACC1) as a target of YB-1. Depletion of YB-1 markedly decreased MACC1 promoter activity and suppressed the MACC1/c-Met signaling pathway in lung adenocarcinoma cells. Additionally, chromatin immunoprecipitation (ChIP) assay demonstrated that YB-1 bound to the MACC1 promoter. Moreover, YB-1 was positively correlated with MACC1, and both proteins were over-expressed in lung adenocarcinoma tissues. The Cox-regression analysis indicated that high YB-1 expression was an independent risk factor for prognosis in enrolled patients. Furthermore, depletion of YB-1 attenuated tumorigenesis in a xenograft mouse model and reduced MACC1 expression in tumor tissues. Collectively, our data suggested that targeting YB-1 suppressed lung adenocarcinoma progression through the MACC1/c-Met pathway and that the high expression of YB-1/MACC1 is a potential prognostic marker in lung adenocarcinoma.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid