Merck
  • Ketohexokinase C blockade ameliorates fructose-induced metabolic dysfunction in fructose-sensitive mice.
Ketohexokinase C blockade ameliorates fructose-induced metabolic dysfunction in fructose-sensitive mice.
The Journal of clinical investigation (2018-03-14)
Miguel A Lanaspa, Ana Andres-Hernando, David J Orlicky, Christina Cicerchi, Cholsoon Jang, Nanxing Li, Tamara Milagres, Masanari Kuwabara, Michael F Wempe, Joshua D Rabinowitz, Richard J Johnson, Dean R Tolan
ABSTRACT
Increasing evidence suggests a role for excessive intake of fructose in the Western diet as a contributor to the current epidemics of metabolic syndrome and obesity. Hereditary fructose intolerance (HFI) is a difficult and potentially lethal orphan disease associated with impaired fructose metabolism. In HFI, the deficiency of aldolase B results in the accumulation of intracellular phosphorylated fructose, leading to phosphate sequestration and depletion, increased adenosine triphosphate (ATP) turnover, and a plethora of conditions that lead to clinical manifestations such as fatty liver, hyperuricemia, Fanconi syndrome, and severe hypoglycemia. Unfortunately, there is currently no treatment for HFI, and avoiding sugar and fructose has become challenging in our society. In this report, through use of genetically modified mice and pharmacological inhibitors, we demonstrate that the absence or inhibition of ketohexokinase (Khk), an enzyme upstream of aldolase B, is sufficient to prevent hypoglycemia and liver and intestinal injury associated with HFI. Herein we provide evidence for the first time to our knowledge of a potential therapeutic approach for HFI. Mechanistically, our studies suggest that it is the inhibition of the Khk C isoform, not the A isoform, that protects animals from HFI.