Merck
  • PAWS1 controls cytoskeletal dynamics and cell migration through association with the SH3 adaptor CD2AP.
PAWS1 controls cytoskeletal dynamics and cell migration through association with the SH3 adaptor CD2AP.
Journal of cell science (2017-11-28)
Timothy D Cummins, Kevin Z L Wu, Polyxeni Bozatzi, Kevin S Dingwell, Thomas J Macartney, Nicola T Wood, Joby Varghese, Robert Gourlay, David G Campbell, Alan Prescott, Eric Griffis, James C Smith, Gopal P Sapkota
ABSTRACT
Our previous studies of PAWS1 (protein associated with SMAD1; also known as FAM83G) have suggested that this molecule has roles beyond BMP signalling. To investigate these roles, we have used CRISPR/Cas9 to generate PAWS1-knockout U2OS osteosarcoma cells. Here, we show that PAWS1 plays a role in the regulation of the cytoskeletal machinery, including actin and focal adhesion dynamics, and cell migration. Confocal microscopy and live cell imaging of actin in U2OS cells indicate that PAWS1 is also involved in cytoskeletal dynamics and organization. Loss of PAWS1 causes severe defects in F-actin organization and distribution as well as in lamellipodial organization, resulting in impaired cell migration. PAWS1 interacts in a dynamic fashion with the actin/cytoskeletal regulator CD2AP at lamellae, suggesting that its association with CD2AP controls actin organization and cellular migration. Genetic ablation of CD2AP from U2OS cells instigates actin and cell migration defects reminiscent of those seen in PAWS1-knockout cells.This article has an associated First Person interview with the first authors of the paper.
Sigma-Aldrich
Anti-CD2AP Antibody, clone 2A2.1, clone 2A2.1, from mouse