Merck
  • Differential NF2 Gene Status in Sporadic Vestibular Schwannomas and its Prognostic Impact on Tumour Growth Patterns.
Differential NF2 Gene Status in Sporadic Vestibular Schwannomas and its Prognostic Impact on Tumour Growth Patterns.
Scientific reports (2017-07-16)
Hongsai Chen, Lu Xue, Hantao Wang, Zhaoyan Wang, Hao Wu
ABSTRACT
The great majority of sporadic vestibular schwannomas (VSs) are due to the inactivation of the NF2 gene. In this study, we found age-dependent differences in the clinical parameters of sporadic VSs. Young patients were characterized by progressive tumour behaviours, including earlier onset of initial symptoms, shorter symptom duration and larger tumour size. An increased rate of "two-hits" of both NF2 alleles, usually by mutation and allelic loss, was observed in young cases compared to older, and this correlated with the loss of protein and mRNA expression. In contrast, the tumours with a single mutation (referred to as 'one-hit') exhibited obvious expression levels. Moreover, a mixture of merlin-expressing tumour cells and non-expressing tumour cells was observed in 'one-hit' schwannomas, suggesting that a subset of 'one-hit' tumour cells was present in these tumours. To mimic the growth promoting effects by the second hit, we performed lentivirus-mediated NF2 knockdown in the 'one-hit' schwannoma cultures. Following the loss of NF2 expression, schwannoma cultures demonstrated increased proliferation rates. Above all, we have identified a correlation between the NF2 status and the growth patterns of sporadic VSs. The treatment decision-making, microsurgery or "wait and scan" strategy, should be carried out according to the tumour's genetic background.