Merck
  • In vitro and in vivo characterization of a novel Hedgehog signaling antagonist in human glioblastoma cell lines.
In vitro and in vivo characterization of a novel Hedgehog signaling antagonist in human glioblastoma cell lines.
International journal of cancer (2011-11-11)
Pietro Ferruzzi, Federica Mennillo, Antonella De Rosa, Cinzia Giordano, Marco Rossi, Giovanni Benedetti, Roberta Magrini, Gal la Pericot Mohr, Vincenzo Miragliotta, Letizia Magnoni, Elisa Mori, Russell Thomas, Patrizia Tunici, Annette Bakker
ABSTRACT
Glioblastoma multiforme (GBM) is composed of heterogeneous and genetically different cells, which are highly invasive and motile. The standard chemotherapeutic agent, temozolomide, affects GBM cell proliferation but is generally unable to prevent tumor recurrence. Hedgehog pathway activation has been reported to be relevant in GBM and different pharmacological pathway modulators have been identified. We report that by growing a commercially available recurrent GBM cell line (DBTRG-05MG) without serum and in the presence of defined growth factors; we obtained a less differentiated cell population, growing in suspension as neurospheres, in which the Hedgehog pathway is activated. Furthermore, the expression profile of Hedgehog pathway components found in DBTRG-05MG neurospheres is similar to primary stem-like cells derived from recurrent GBM patients. We report the effect of our novel specific Smoothened receptor antagonist (SEN450) on neurosphere growing cells and compared its effect to that of well known benchmark compounds. Finally, we showed that SEN450 is both antiproliferative on its own and further reduces tumor volume after temozolomide pretreatment in a mouse xenograft model using DBTRG-05MG neurosphere cells. Altogether our data indicate that the Hedgehog pathway is not irreversibly switched off in adherent cells but can be reactivated when exposed to well-defined culture conditions, thus restoring the condition observed in primary tumor-derived material, and that pharmacological modulation of this pathway can have profound influences on tumor proliferation. Therefore, pharmacological inhibition of the Hedgehog pathway is a potentially useful therapeutic approach in GBM.
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture