Merck
  • Clonogenic analysis reveals reserve stem cells in postnatal mammals. II. Pluripotent epiblastic-like stem cells.
Clonogenic analysis reveals reserve stem cells in postnatal mammals. II. Pluripotent epiblastic-like stem cells.
The anatomical record. Part A, Discoveries in molecular, cellular, and evolutionary biology (2004-02-26)
Henry E Young, Cecile Duplaa, Michael J Yost, Nicholas L Henson, Julie A Floyd, Kristina Detmer, Angela J Thompson, Steven W Powell, T Clark Gamblin, Kirk Kizziah, Benjamin J Holland, Angel Boev, J M Van De Water, Dan C Godbee, Stephanie Jackson, Marylen Rimando, Chad R Edwards, Eveline Wu, Chris Cawley, Pamela D Edwards, Anna Macgregor, Ryan Bozof, T Michele Thompson, George J Petro, Heather M Shelton, Beth L McCampbell, Jared C Mills, Frederick L Flynt, Timothy A Steele, Marianne Kearney, Amy Kirincich-Greathead, Wade Hardy, Paul R Young, Aman V Amin, R Steve Williams, Miranda M Horton, Shaun McGuinn, Kristina C Hawkins, Kurt Ericson, Louis Terracio, Catherine Moreau, Douglas Hixson, Brian W Tobin, John Hudson, Frank P Bowyer, Asa C Black
ABSTRACT
Undifferentiated cells have been identified in the prenatal blastocyst, inner cell mass, and gonadal ridges of rodents and primates, including humans. After isolation these cells express molecular and immunological markers for embryonic cells, capabilities for extended self-renewal, and telomerase activity. When allowed to differentiate, embryonic stem cells express phenotypic markers for tissues of ectodermal, mesodermal, and endodermal origin. When implanted in vivo, undifferentiated noninduced embryonic stem cells formed teratomas. In this report we describe a cell clone isolated from postnatal rat skeletal muscle and derived by repetitive single-cell clonogenic analysis. In the undifferentiated state it consists of very small cells having a high ratio of nucleus to cytoplasm. The clone expresses molecular and immunological markers for embryonic stem cells. It exhibits telomerase activity, which is consistent with its extended capability for self-renewal. When induced to differentiate, it expressed phenotypic markers for tissues of ectodermal, mesodermal, and endodermal origin. The clone was designated as a postnatal pluripotent epiblastic-like stem cell (PPELSC). The undifferentiated clone was transfected with a genomic marker and assayed for alterations in stem cell characteristics. No alterations were noted. The labeled clone, when implanted into heart after injury, incorporated into myocardial tissues undergoing repair. The labeled clone was subjected to directed lineage induction in vitro, resulting in the formation of islet-like structures (ILSs) that secreted insulin in response to a glucose challenge. This study suggests that embryonic-like stem cells are retained within postnatal mammals and have the potential for use in gene therapy and tissue engineering.
Sigma-Aldrich
Anti-Myosin Antibody, heavy chain β, culture supernatant, clone 5B9 (2C8), Chemicon®
Sigma-Aldrich
Anti-Nestin Antibody, clone rat-401, clone rat-401, Chemicon®, from mouse