Merck
  • Mechanistic studies of the toxicity of zinc gluconate in the olfactory neuronal cell line Odora.
Mechanistic studies of the toxicity of zinc gluconate in the olfactory neuronal cell line Odora.
Toxicology in vitro : an international journal published in association with BIBRA (2016-05-18)
Heidi Hsieh, Kavitha Subramanian Vignesh, George S Deepe, Divaker Choubey, Howard G Shertzer, Mary Beth Genter
ABSTRACT
Zinc is both an essential and potentially toxic metal. It is widely believed that oral zinc supplementation can reduce the effects of the common cold; however, there is strong clinical evidence that intranasal (IN) zinc gluconate (ZG) gel treatment for this purpose causes anosmia, or the loss of the sense of smell, in humans. Using the rat olfactory neuron cell line, Odora, we investigated the molecular mechanism by which zinc exposure exerts its toxic effects on olfactory neurons. Following treatment of Odora cells with 100 and 200μM ZG for 0-24h, RNA-seq and in silico analyses revealed up-regulation of pathways associated with zinc metal response, oxidative stress, and ATP production. We observed that Odora cells recovered from zinc-induced oxidative stress, but ATP depletion persisted with longer exposure to ZG. ZG exposure increased levels of NLRP3 and IL-1β protein levels in a time-dependent manner, suggesting that zinc exposure may cause an inflammasome-mediated cell death, pyroptosis, in olfactory neurons.