Merck
  • Critical role of mast cell chymase in mouse abdominal aortic aneurysm formation.
Critical role of mast cell chymase in mouse abdominal aortic aneurysm formation.
Circulation (2009-09-02)
Jiusong Sun, Jie Zhang, Jes S Lindholt, Galina K Sukhova, Jian Liu, Aina He, Magnus Abrink, Gunnar Pejler, Richard L Stevens, Robert W Thompson, Terri L Ennis, Michael F Gurish, Peter Libby, Guo-Ping Shi
ABSTRACT
Mast cell chymase may participate in the pathogenesis of human abdominal aortic aneurysm (AAA), yet a direct contribution of this serine protease to AAA formation remains unknown. Human AAA lesions had high numbers of chymase-immunoreactive mast cells. Serum chymase level correlated with AAA growth rate (P=0.009) in a prospective clinical study. In experimental AAA produced by aortic elastase perfusion in wild-type (WT) mice or those deficient in the chymase ortholog mouse mast cell protease-4 (mMCP-4) or deficient in mMCP-5 (Mcpt4(-/-), Mcpt5(-/-)), Mcpt4(-/-) but not Mcpt5(-/-) had reduced AAA formation 14 days after elastase perfusion. Even 8 weeks after perfusion, aortic expansion in Mcpt4(-/-) mice fell by 50% compared with that of the WT mice (P=0.0003). AAA lesions in Mcpt4(-/-) mice had fewer inflammatory cells and less apoptosis, angiogenesis, and elastin fragmentation than those of WT mice. Although Kit(W-sh/W-sh) mice had protection from AAA formation, reconstitution with mast cells from WT mice, but not those from Mcpt4(-/-) mice, partially restored the AAA phenotype. Mechanistic studies suggested that mMCP-4 regulates expression and activation of cysteine protease cathepsins, elastin degradation, angiogenesis, and vascular cell apoptosis. High chymase-positive mast cell content in human AAA lesions, greatly reduced AAA formation in Mcpt4(-/-) mice, and significant correlation of serum chymase levels with human AAA expansion rate suggests participation of mast cell chymase in the progression of human and mouse AAA.
Sigma-Aldrich
Anti-Chymase Antibody, clone B7, biotin conjugated, clone B7, Chemicon®, from mouse