Merck
  • Involvement of cAMP in nerve growth factor-triggered p35/Cdk5 activation and differentiation in PC12 cells.
Involvement of cAMP in nerve growth factor-triggered p35/Cdk5 activation and differentiation in PC12 cells.
American journal of physiology. Cell physiology (2010-05-14)
Mei-Chih Chen, Ho Lin, Fu-Ning Hsu, Pao-Hsuan Huang, Guan-Shun Lee, Paulus S Wang
ABSTRACT
The signaling mechanisms underlying cell differentiation have been extensively studied with the use of rat PC12 cells as a model system. Nerve growth factor (NGF) is a trophic factor inducing PC12 cell differentiation through the activation of the p35/cyclin-dependent kinase 5 (Cdk5) complex. It has been reported that adenylyl cyclase activation and cAMP production may be involved in NGF-dependent actions. Our previous results indicate that cAMP activates the p35/Cdk5 complex in reproductive cells. Therefore, the role of cAMP in NGF-triggered p35/Cdk5 activation and PC12 differentiation was interesting to explore. Our results indicate that roscovitine, a molecular inhibitor of Cdk5, blocks cAMP-triggered PC12 differentiation, which was evaluated by neurite initiation, a decrease in proliferation, and cell cycle G(1) arrest. The following data show that cAMP treatment increased Cdk5 activity through p35 upregulation. cAMP downstream components, protein kinase A (PKA) and phosphorylated cAMP response element binding protein (CREB), are involved in this regulation. The immunocytochemical results indicate that PKA inhibition disrupted cAMP-triggered p35/Cdk5 localization in PC12 cells. In addition, adenylyl cyclase inhibition was found to diminish NGF-induced intracellular cAMP production, CREB phosphorylation, and p35 expression. The cAMP antagonist and the PKA inhibitors reduced NGF-induced p35 expression. Finally, NGF-triggered PC12 differentiation was partially decreased by adenylyl cyclase or PKA inhibitors. In conclusion, these results demonstrate that cAMP may play a role in NGF-p35/Cdk5-dependent PC12 differentiation.
Sigma-Aldrich
KT 5720, ≥98% (HPLC), powder
Sigma-Aldrich
H-89 dihydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
2′,5′-Dideoxyadenosine, ≥95% (HPLC), solid
Sigma-Aldrich
8-Bromoadenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
Roscovitine, ≥98% (TLC)
Sigma-Aldrich
SQ 22,536, ≥97% (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
Histone H1 Protein, 20 mg, Purified bovine Histone H1 protein for use as a substrate in histone modification assays (HAT, HDAC, DNMT) and chromatin assembly studies.