Merck
  • Alterations in cognitive outcome between 3 and 12 months in survivors of out-of-hospital cardiac arrest.
Alterations in cognitive outcome between 3 and 12 months in survivors of out-of-hospital cardiac arrest.
Resuscitation (2016-06-04)
Marte Ørbo, Per M Aslaksen, Kristina Larsby, Christoph Schäfer, Pål M Tande, Audny Anke
ABSTRACT
To prospectively investigate cognitive recovery from 3 to 12 months after resuscitation from out-of-hospital cardiac arrest (OHCA) and the associations between cognitive performance at 3 months and health-related quality of life (HRQL), psychological distress and work status after 12 months. At both assessments, neuropsychological tests were used to measure aspects of general mental ability, verbal and visual memory, psychomotor speed and executive function. The Short Form-36 (SF-36) was used to measure mental and physical HRQL, and the Hospital Anxiety and Depression Scale (HADS) to assess psychological distress. 33 survivors completed both exams (31 males, mean age 58.6 years, SD=13). The OHCAs were witnessed and due to cardiac origins. Nine patients were awake at admission to the hospital. Longer coma duration was associated with poorer cognitive results. Memory impairments were the most common symptom. The mean changes and effect sizes indicated minor improvements in cognitive performance from 3 to 12 months (Hedges g≤.26). Reliable change indices for an individual's results further confirmed the stability of the group statistics. The HADS scores showed increased depressive symptoms, and mental HRQL was reduced from 3 to 12 months. Higher reports of psychological distress were related to worse HRQL. Work participation increased. Better cognitive results at 3 months were correlated with better HRQL and return to work at 12 months. The current data describe stability in results from 3 to 12 months. A worse cognitive performance at 3 months and higher reports of psychological distress were associated with lower HRQL.
Product Description
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)