Merck
  • The role of pi-acidic and pi-basic chiral stationary phases in the high-performance liquid chromatographic enantioseparation of unusual beta-amino acids.
The role of pi-acidic and pi-basic chiral stationary phases in the high-performance liquid chromatographic enantioseparation of unusual beta-amino acids.
Chirality (2008-06-17)
István Ilisz, Róbert Berkecz, Eniko Forró, Ferenc Fülöp, Daniel W Armstrong, Antal Péter
ABSTRACT
The application of 3,5-dimethylphenyl-carbamoylated-beta-cyclodextrin (Cyclobond I 2000 DMP) and 2,6-dinitro-4-trifluoromethylphenyl-ether-beta-cyclodextrin-based (Cyclobond DNP) chiral stationary phases for the high-performance liquid chromatographic enantioseparation of unusual beta-amino acids is reported. The investigated amino acids were saturated or unsaturated alicyclic beta-3-homo-amino acids and bicyclic beta-amino acids. Prior to chromatographic analyses, all amino acids were transformed to N-3,5-dinitrobenzoyl- or N-3,5-dimethylbenzoyl form to ensure a pi-acidic or pi-basic function and to enhance the pi-acidic-pi-basic interactions between analytes and chiral selectors. Chromatographic results are given as retention, separation and resolution factors. The chromatographic conditions were varied to achieve optimal separation. The sequence of elution of the enantiomers was determined in some cases.
Product Description
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CYCLOBOND I 2000 DMP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CYCLOBOND I 2000 DMP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CYCLOBOND I 2000 DMP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm