Merck
HomeSupportCalculators & AppsWire Gauge Conversion Chart
Wire Gauge Conversion Chart
Standard Wire Gauge Chart for AWG Conversion
Use this Standard Wire Gauge Chart for simple conversions and reference while you work. Convert standardized AWG (American Wire Gauge) sizes to inches (in) and millimeters (mm).
Download Wire Gauge PDF
Gauge No. (AWG)InchesMillimeters
0000000.580014.73
000000.516513.12
00000.460011.68
0000.409610.40
000.36489.266
00.32498.251
10.28937.348
20.25766.544
30.22945.827
40.20435.189
50.18194.621
60.16204.115
70.14433.665
80.12853.264
90.11442.906
100.10192.588
110.090742.305
120.080812.053
130.071961.828
140.064081.628
150.057071.45
160.050821.291
170.045261.149
180.040301.024
190.035890.9116
200.031960.8118
210.028460.7229
220.025350.6438
230.022570.5733
240.020100.5106
250.017900.4547
260.015940.4049
270.014200.3606
280.012640.3211
290.011260.2859
300.010020.2546
310.0089280.2268
320.0079500.2019
330.0070800.1798
340.0063050.1601
350.0056150.1426
360.0050000.127
370.0044530.1131
380.0039650.1007
390.0035300.08969
400.0031440.07986