Skip to Content
Merck
Zirconium(IV) tert-butoxide electronic grade, 99.999% trace metals basis

Zirconium(IV) tert-butoxide

Zirconium(IV) tert-butoxide electronic grade, 99.999% trace metals basis
Synonym(s):
Tetra-tert-butyl zirconate
Linear Formula:
Zr[OC(CH3)3]4
CAS Number:
Molecular Weight:
383.68

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing