Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Zinc sulfide
Products
Manufacturer
Physical Form
Particle Size (µm)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
GF41490794
Zinc sulfide,
powder, max. particle size 45 micron, weight 100 g, purity 99.9%
Expand
View Pricing
GF13437031
Zinc sulfide,
powder, max. particle size 45 micron, weight 50 g, purity 99.9%
Expand
View Pricing
GF75209171
Zinc sulfide,
lump, 5 mm max. lump size, weight 100 g, purity 99.99%
Expand
View Pricing
GF17917713
Zinc sulfide,
powder, max. particle size 45 micron, weight 200 g, purity 99.9%
Expand
View Pricing
GF08737083
Zinc sulfide,
lump, 5 mm max. lump size, weight 50 g, purity 99.99%
Expand
View Pricing
GF27902347
Zinc sulfide,
lump, 5 mm max. lump size, weight 20 g, purity 99.99%
Expand
View Pricing
GF79586803
Zinc sulfide,
powder, 45 max. part. size (micron), weight 500 g, purity 99.9%
Expand
View Pricing