Skip to Content
Products
Cart
0
AR
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Zinc acetate dihydrate
Linear Formula:
Zn(CH
3
COO)
2
· 2H
2
O
CAS Number:
5970-45-6
Molecular Weight:
219.51
EC Number:
209-170-2
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (5)
Formula Weight
pH Value
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
ACS reagent (2)
Puriss (2)
Puriss P.A. (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
383058
Zinc acetate dihydrate,
ACS reagent, ≥98%
Expand
View Pricing
96459
Zinc acetate dihydrate,
puriss. p.a., ACS reagent, ≥99.0% (KT)
Expand
View Pricing
Z0625
Zinc acetate dihydrate,
reagent grade
Expand
View Pricing
379786
Zinc acetate dihydrate,
99.999% trace metals basis
Expand
View Pricing
25044
Zinc acetate dihydrate,
puriss., E 650, 99-102%
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
Español - ES
English - EN
Learn More