Skip to Content
Merck
Zenix® SEC-300 Gel Filtration Column, 3 μm L × I.D. 25 cm × 4.6 mm, 3 μm particle size, stainless steel

Zenix® SEC-300 (3 µm) HPLC Columns

Zenix® SEC-300 Gel Filtration Column, 3 μm L × I.D. 25 cm × 4.6 mm, 3 μm particle size, stainless steel

Products

Product Category
Manufacturer
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-15 of 15
Compare
Product Number
Description
Pricing